missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-dimethoxybenzene, 98%

Product Code. 11491050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491050 1 g 1g
11401060 5 g 5g
11411060 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491050 Supplier Thermo Scientific Alfa Aesar Supplier No. B25485.03
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 16932-45-9 | C8H9BrO2 | 217.062 g/mol

2-Bromo-1,3-dimethoxybenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,3-dimethoxybenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16932-45-9
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.062
MDL Number MFCD07780177
InChI Key VHVYSMMZHORFKU-UHFFFAOYSA-N
PubChem CID 612296
IUPAC Name 2-bromo-1,3-dimethoxybenzene
SMILES COC1=C(C(=CC=C1)OC)Br

Specifications

Melting Point 91°C to 94°C
Quantity 1 g
Synonym 1-bromo-2,6-dimethoxybenzene, 2-bromo-3-methoxyanisole, 2,6-dimethoxybromobenzene, benzene, bromo-1,3-dimethoxy, benzene,2-bromo-1,3-dimethoxy, 1,3-dimethoxy-2-bromobenzene, acmc-209dzi, 2.6-dimethoxybromobenzene, benzene, 2-bromo-1,3-dimethoxy
Solubility Information Insoluble in water.
Formula Weight 217.06
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,3-dimethoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.