missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3-ethyl-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one, ≥95%, Thermo Scientific™

Product Code. 10582852
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10582852 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10582852 Supplier Thermo Scientific Maybridge Supplier No. CD00361DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 175136-57-9
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C18H25BrO
MDL Number MFCD00052015
Quantity 1 g
Synonym 2-bromo-1-3-ethyl-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 7-bromoacetyl-6-ethyl-1,2,3,4-tetrahydro-1,1,4,4-tetramethylnaphthalene, 2-bromo-1-3-ethyl-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethanone, 2-bromo-1-3-ethyl-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one, 2-bromanyl-1-3-ethyl-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethanone
InChI Key BBHJYKJAWKZSHS-UHFFFAOYSA-N
SMILES CCC1=CC2=C(C=C1C(=O)CBr)C(CCC2(C)C)(C)C
IUPAC Name 2-bromo-1-(3-ethyl-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethanone
Molecular Weight (g/mol) 337.301
PubChem CID 254968
Formula Weight 337.3
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(3-ethyl-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.