missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,4-dimethoxybenzene, 98%

Product Code. 11407547
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11407547

Brand: Thermo Scientific Alfa Aesar A15640.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25245-34-5 | C8H9BrO2 | 217.06 g/mol

2-Bromo-1,4-dimethoxybenzene has been used in preparation of S-(-)-2-[N-(trifluoroacetyl)amino]-1-(2,5-dimethoxy-4- bromophenyl)-1-propanone, 1,4-bis{4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1,4-bis{2,5-dimethoxy-4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1-bromo-4-iodo-2,5-dimethoxybenzene. It can be used as a chemical redox shuttle additive in Lithium ion batteries for repeated overcharge and overdischarge protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,4-dimethoxybenzene has been used in preparation of S-(-)-2-[N-(trifluoroacetyl)amino]-1-(2,5-dimethoxy-4- bromophenyl)-1-propanone, 1,4-bis{4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1,4-bis{2,5-dimethoxy-4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1-bromo-4-iodo-2,5-dimethoxybenzene. It can be used as a chemical redox shuttle additive in Lithium ion batteries for repeated overcharge and overdischarge protection.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25245-34-5
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD00008355
InChI Key DWCGNRKFLRLWCJ-UHFFFAOYSA-N
PubChem CID 91353
IUPAC Name 2-bromo-1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C(Br)=C1

Specifications

Density 1.441
Boiling Point 260°C to 262°C
Flash Point >110°C (230°F)
Refractive Index 1.57
Quantity 10 g
Beilstein 2441884
Synonym 1-bromo-2,5-dimethoxybenzene, benzene, 2-bromo-1,4-dimethoxy, 2,5-dimethoxybromobenzene, pubchem19785, 2-bromo-4-methoxyanisole, acmc-1cby3, ksc490m9p, 1,4-dimethoxy-2-bromobenzene, 1-bromo-2, 5-dimethoxybenzene, dwcgnrkflrlwcj-uhfffaoysa
Solubility Information Insoluble in water.
Formula Weight 217.07
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,4-dimethoxybenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.