missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-phenoxyphenyl)ethanone 95+%, Thermo Scientific™

Product Code. 10703907
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10703907 5 g 5g
10032464 250 mg 250mg
10282933 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10703907 Supplier Thermo Scientific Maybridge Supplier No. CC53705DE
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. Se returpolitik

Kemiske identifikatorer

CAS 28179-33-1
Molecular Formula C14H11BrO2
Molecular Weight (g/mol) 291.14
MDL Number MFCD01419855
InChI Key RAXTYMXDSNWNJS-UHFFFAOYSA-N
Synonym 2-bromo-1-4-phenoxyphenyl ethanone, 2-bromo-1-4-phenoxyphenyl ethan-1-one, ethanone,2-bromo-1-4-phenoxyphenyl, 4-phenoxyphenacyl bromide, 2-bromo-4'-phenoxyacetophenone, 4'-phenoxy-2-bromoacetophenone, alpha-bromo-4'-phenoxyacetophenone, ethanone, 2-bromo-1-4-phenoxyphenyl
PubChem CID 14797675
IUPAC Name 2-bromo-1-(4-phenoxyphenyl)ethanone
SMILES BrCC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Tekniske data

Assay Percent Range 90%
Quantity 5 g
Formula Weight 291.14
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(4-phenoxyphenyl)ethanone
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.