missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10385102
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10385102 250 mg Amber Glass Bottle 250mg
10611123 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10385102 Supplier Thermo Scientific Maybridge Supplier No. TL00641CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 132392-28-0
Molecular Formula C16H21BrO
Molecular Weight (g/mol) 309.25
MDL Number MFCD00178768
InChI Key BWXPCWLVNVLIFR-UHFFFAOYSA-N
Synonym 2-bromo-1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one, 6-bromoacetyl-1,2,3,4-tetrahydro-1,1,4,4-tetramethylnaphthalene, 2-bromo-1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 2-bromo-1-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethanone, 2-bromo-1-5,5,8,8-tetramethyl 2-5,6,7,8-tetrahydronaphthyl ethan-1-one, 2-bromo-1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-on e
PubChem CID 2801337
IUPAC Name 2-bromo-1-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethanone
SMILES CC1(C)CCC(C)(C)C2=C1C=CC(=C2)C(=O)CBr

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 309.24
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.