missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3,4-difluoroaniline, 97%

Product Code. 15458468
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458468 25 g 25g
15438468 1 g 1g
15448468 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458468 Supplier Thermo Scientific Alfa Aesar Supplier No. H61876.14
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1092349-87-5 | C6H4BrF2N | 208.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092349-87-5
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.01
MDL Number MFCD11035924
InChI Key ZGZURPPCPVPDMX-UHFFFAOYSA-N
Synonym 2-bromo-3,4-difluoro aniline, 2-bromo-3,4-difluorophenylamine, 2-bromo-3,4-difluorobenzenamine
PubChem CID 22731604
IUPAC Name 2-bromo-3,4-difluoroaniline
SMILES NC1=C(Br)C(F)=C(F)C=C1

Specifications

Quantity 25 g
UN Number UN2811
Formula Weight 208
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromo-3,4-difluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.