missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoroaniline, 97%

Product Code. 15498258
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498258 5 g 5g
15408268 250 mg 250mg
15488258 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498258 Supplier Thermo Scientific Alfa Aesar Supplier No. H61687.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111721-75-6 | C6H5BrFN | 190.015 g/mol

2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111721-75-6
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD07369915
InChI Key XZRSXRUYZXBTGD-UHFFFAOYSA-N
Synonym 2-bromo-3-fluorophenylamine, 2-bromo-3-fluoro-phenylamine, 2-bromo-3-fluorobenzenamine, benzenamine, 2-bromo-3-fluoro, pubchem1523, acmc-1c6kb, 2-bromo-3-fluoro-aniline, ksc493i1t, 2-bromo-3-fluoro-phenyl-amine
PubChem CID 14770005
IUPAC Name 2-bromo-3-fluoroaniline
SMILES C1=CC(=C(C(=C1)F)Br)N

Specifications

Color White
Density 1.670 g/mL
Flash Point 94°C (201°F)
Refractive Index 1.583
Quantity 5 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 190.01
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 2-Bromo-3-fluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.