missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylaniline, 97+%

Product Code. p-7055436
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15461029

Brand: Thermo Scientific Alfa Aesar H64014.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54879-20-8 | C7H8BrN | 186.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54879-20-8
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD06411367
InChI Key VJNUZLYTGSGDHR-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-3-methyl, 3-amino-2-bromotoluene, 2-bromo-3-methylbenzenamine, 2-bromo-3-methyl-benzenamine, 2-bromo-3-methylphenylamine, 2-bromo-3-methyl-phenylamine, acmc-20af7i, 2-bromo-3-methyl aniline, ksc602o1n, benzenamine,2-bromo-3-methyl
PubChem CID 603781
IUPAC Name 2-bromo-3-methylaniline
SMILES CC1=C(C(=CC=C1)N)Br

Specifications

Color Brown
Density 1.489 g/mL
Boiling Point 48°C (0.03 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.605
Quantity 250 mg
UN Number UN2810
Beilstein 2079474
Formula Weight 186.05
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-methylaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.