missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals

Product Code. 15451459
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451459 5 g 5g
15441459 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15451459 Supplier Thermo Scientific Alfa Aesar Supplier No. H64309.06
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58458-10-9
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.023
MDL Number MFCD01656033
InChI Key XIUJCAJFRWDEKE-UHFFFAOYSA-N
Synonym 2-bromo-3-trifluoromethyl aniline, 3-amino-2-bromobenzotrifluoride, 2-bromo-3-trifluoromethyl benzenamine, 2-bromo-3-trifluoromethyl phenylamine, benzenamine, 2-bromo-3-trifluoromethyl, pubchem4829, ksc275a9t, 2-bromo-3-trifluoromethyl-aniline, 3-amino-2-bromobenzotrifluoride 2-bromo-3-trifluoromethyl aniline
PubChem CID 94058
IUPAC Name 2-bromo-3-(trifluoromethyl)aniline
SMILES C1=CC(=C(C(=C1)N)Br)C(F)(F)F

Specifications

Color White
Quantity 5 g
UN Number UN2811
Beilstein 2835356
Formula Weight 240.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.