missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-dichloroaniline, 98%, Thermo Scientific™

Product Code. 10756085 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10756085 5 g Glass bottle 5g
10766085 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10756085 Supplier Thermo Scientific Acros Supplier No. 399600050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 697-86-9
Molecular Formula C6H4BrCl2N
Molecular Weight (g/mol) 240.91
MDL Number MFCD00040936
InChI Key DTPADCOGQUOGHT-UHFFFAOYSA-N
Synonym pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline
PubChem CID 2756901
IUPAC Name 2-bromo-4,6-dichloroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Br)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 80.0°C to 81.0°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H2(Br)NH2
Quantity 5 g
Formula Weight 240.91
Percent Purity 98%
Chemical Name or Material 2-Bromo-4, 6-dichloroaniline
compliance-icons
Product Identifier
  • 2-Bromo-4,6-dichloroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.