missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-difluoroaniline, 98%

Product Code. 11458853
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458853 500 g 500g
11438853 25 g 25g
11448853 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458853 Supplier Thermo Scientific Alfa Aesar Supplier No. A11022.36
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 444-14-4 | C6H4BrF2N | 208.01 g/mol

2-Bromo-4,6-difluoroaniline is used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,6-difluoroaniline is used in chemical synthesis.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444-14-4
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.01
MDL Number MFCD00009639
InChI Key WUJKFVGKLTWVSQ-UHFFFAOYSA-N
Synonym 2,4-difluoro-6-bromoaniline, 6-bromo-2,4-difluoroaniline, 2-bromo-4,6-difluoro-phenylamine, 2-bromo-4,6-difluorobenzenamine, benzenamine, 2-bromo-4,6-difluoro, 2-bromo-4,6-difluorophenylamine, pubchem2913, acmc-1ao2h, 2-bromo4,6-difluoroaniline
PubChem CID 136285
IUPAC Name 2-bromo-4,6-difluoroaniline
SMILES NC1=C(F)C=C(F)C=C1Br

Specifications

Melting Point 38°C to 42°C
Odor Weak, characteristic
Quantity 500 g
UN Number UN3077
Beilstein 2718139
Solubility Information Insoluble in water.
Formula Weight 208.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4,6-difluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.