missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloro-1-iodobenzene, 98%

Product Code. 13551630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13551630 25 g Glass bottle 25g
13531630 1 g Glass bottle 1g
13541630 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13551630 Supplier Thermo Scientific Acros Supplier No. 448760250
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31928-44-6 | C6H3BrClI | 317.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31928-44-6
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.36
InChI Key CXHXFDQEFKFYQJ-UHFFFAOYSA-N
Synonym 2-bromo-4-chloroiodobenzene, benzene, 2-bromo-4-chloro-1-iodo, acmc-209hpo, ksc494q6f, 1-iodo-2-bromo-4-chlorobenzene, 2-bromo-1-iodo-4-chlorobenzene
PubChem CID 13741326
IUPAC Name 2-bromo-4-chloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Cl)Br)I

Spécification

Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(Cl)I
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 317.36
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-chloro-1-iodobenzene
compliance-icons
Identifiant produit
  • 2-Bromo-4-chloro-1-iodobenzene
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.