missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chlorophenol, 98+%

Product Code. 11443317
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11443317

Brand: Thermo Scientific Alfa Aesar A16218.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 695-96-5 | C6H4BrClO | 207.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-96-5
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.45
MDL Number MFCD00002319
InChI Key ZIYRDJLAJYTELF-UHFFFAOYSA-N
Synonym phenol, 2-bromo-4-chloro, 4-chloro-2-bromophenol, 2-bromo-4-chloro-phenol, 4-chloro-2-bromo phenol, pubchem3594, 2-bromo-4-chorophenol, acmc-1ayt0, akos bbb/602, ksc493o9n, 2-bromo-4-chlorophenol
PubChem CID 69670
IUPAC Name 2-bromo-4-chlorophenol
SMILES OC1=CC=C(Cl)C=C1Br

Specifications

Melting Point 31°C to 35°C
Boiling Point 230°C to 232°C
Flash Point >110°C (230°F)
Odor Phenol-like
Quantity 25 g
Beilstein 2042871
Formula Weight 207.46
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-4-chlorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.