missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoro-6-(trifluoromethyl)aniline, 98%

Product Code. 11336629
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11336629 10 g 10g
11326629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336629 Supplier Thermo Scientific Alfa Aesar Supplier No. H26206.09
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 875664-27-0 | C7H4BrF4N | 258.014 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875664-27-0
Molecular Formula C7H4BrF4N
Molecular Weight (g/mol) 258.014
MDL Number MFCD04973756
InChI Key OTYCOVZULSPPPW-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-6-trifluoromethyl aniline, 2-amino-3-bromo-5-fluorobenzotrifluoride, buttpark 154\11-38, benzenamine,2-bromo-4-fluoro-6-trifluoromethyl, 2-bromo-4-fluoro-6-trifluoromethyl benzenamine, pubchem19869, ksc658m8n, 2-bromo-4-fluoro-6-trifluoromethyl phenylamine, 6-bromo-4-fluoro-2-trifluoromethyl phenylamine, 2-bromanyl-4-fluoranyl-6-trifluoromethyl aniline
PubChem CID 7018045
IUPAC Name 2-bromo-4-fluoro-6-(trifluoromethyl)aniline
SMILES C1=C(C=C(C(=C1Br)N)C(F)(F)F)F

Specifications

Refractive Index 1.503
Quantity 10 g
UN Number UN2810
Formula Weight 258.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluoro-6-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.