missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoroanisole, 98%, Thermo Scientific™

Product Code. 10237260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10237260 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10237260 Supplier Thermo Scientific Acros Supplier No. 269800250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 452-08-4
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H6BrFO
Linear Formula BrC6H3(F)OCH3
Refractive Index 1.5440 to 1.5460 (20°C, 589nm)
MDL Number MFCD00012014
Quantity 25 g
Beilstein 06, 1057
Synonym 2-bromo-4-fluoroanisole, benzene, 2-bromo-4-fluoro-1-methoxy, 2-bromo-4-fluoro anisole, 1-bromo-3-fluoro-6-methoxybenzene, 2-bromo-4-fluoro-methoxybenzene, 2-bromo-4-fluoro-1-methoxy-benzene, 1-bromo-5-fluoro-2-methoxybenzene, attercop-chm at106665, pubchem1972, 2-bromo-4-fluoroanisol
InChI Key JIQXVIJARQLCOY-UHFFFAOYSA-N
SMILES COC1=CC=C(F)C=C1Br
IUPAC Name 2-bromo-4-fluoro-1-methoxybenzene
Molecular Weight (g/mol) 205.03
PubChem CID 136292
Formula Weight 205.03
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-fluoroanisole, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.