missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoroanisole, 98%

Product Code. 11468873
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468873 50 g 50g
11458873 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468873 Supplier Thermo Scientific Alfa Aesar Supplier No. A11040.18
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 452-08-4 | C7H6BrFO | 205.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-08-4
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.03
MDL Number MFCD00012014
InChI Key JIQXVIJARQLCOY-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoroanisole, benzene, 2-bromo-4-fluoro-1-methoxy, 2-bromo-4-fluoro anisole, 1-bromo-3-fluoro-6-methoxybenzene, 2-bromo-4-fluoro-methoxybenzene, 2-bromo-4-fluoro-1-methoxy-benzene, 1-bromo-5-fluoro-2-methoxybenzene, attercop-chm at106665, pubchem1972, 2-bromo-4-fluoroanisol
PubChem CID 136292
IUPAC Name 2-bromo-4-fluoro-1-methoxybenzene
SMILES COC1=CC=C(F)C=C1Br

Specifications

Density 1.581
Boiling Point 88°C to 89°C (1 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.545
Quantity 50 g
Beilstein 2358880
Formula Weight 205.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluoroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.