missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzylamine hydrochloride, 96%

Product Code. 15448748
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448748 1 g 1g
15427156 250 mg 250mg
15458748 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448748 Supplier Thermo Scientific Alfa Aesar Supplier No. H62200.03
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 289038-14-8 | C7H8BrClFN | 240.5 g/mol

2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 289038-14-8
Molecular Formula C7H8BrClFN
Molecular Weight (g/mol) 240.5
MDL Number MFCD00672922
InChI Key PBCSNHSXUSMNOV-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzylamine hydrochloride, 2-bromo-4-fluorophenyl methanamine hydrochloride, 1-2-bromo-4-fluorophenyl methanamine hydrochloride, 2-bromanyl-4-fluoranyl-phenyl methanamine hydrochloride
PubChem CID 2773353
IUPAC Name (2-bromo-4-fluorophenyl)methanamine;hydrochloride
SMILES C1=CC(=C(C=C1F)Br)CN.Cl

Specifications

Quantity 1 g
Formula Weight 240.5
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Bromo-4-fluorobenzylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.