missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-isopropylaniline, 97%

Product Code. 11437217
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437217 5 g 5g
11447217 25 g 25g
11457217 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437217 Supplier Thermo Scientific Alfa Aesar Supplier No. A14995.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 51605-97-1 | C9H12BrN | 214.11 g/mol

2-Bromo-4-isopropylaniline is involved in the preparation of 4-(5-aryl-1, 2, 3, 6-tetrahydropyridino) pyrimidine derivatives and 4-substituted-2-anilinopyrimidine derivatives, which find application as corticotropin releasing factor (CRF) antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-isopropylaniline is involved in the preparation of 4-(5-aryl-1, 2, 3, 6-tetrahydropyridino) pyrimidine derivatives and 4-substituted-2-anilinopyrimidine derivatives, which find application as corticotropin releasing factor (CRF) antagonists.

Solubility
Miscible with dioxane and ethoxyethanol. Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51605-97-1
Molecular Formula C9H12BrN
Molecular Weight (g/mol) 214.11
MDL Number MFCD00051585
InChI Key WEMDUNBELVTSRP-UHFFFAOYSA-N
Synonym 2-bromo-4-isopropylaniline, 2-bromo-4-isopropyl-phenylamine, 4-amino-2-bromocumene, benzenamine, 2-bromo-4-1-methylethyl, zlchem 981, pubchem17446, acmc-1anzo, 2-bromo-4-isopropyl aniline, 2-bromo4-isopropyl-phenylamine, 2-bromo-4-2-propyl aniline
PubChem CID 521337
SMILES CC(C)C1=CC=C(N)C(Br)=C1

Specifications

Density 1.35
Boiling Point 116°C to 118°C (3 mmHg)
Quantity 5 g
UN Number UN2810
Beilstein 2638470
Solubility Information Miscible with dioxane and ethoxyethanol. Immiscible with water.
IUPAC Name 2-bromo-4-propan-2-ylaniline
Formula Weight 214.11
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-isopropylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.