missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methoxyphenylacetic acid, 97%

Product Code. 11397459
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397459 1 g 1g
11307469 5 g 5g
11317469 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397459 Supplier Thermo Scientific Alfa Aesar Supplier No. H27859.03
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 66916-99-2 | C9H9BrO3 | 245.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66916-99-2
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD02664700
InChI Key XQELSBAAFMYSMG-UHFFFAOYSA-N
Synonym 2-bromo-4-methoxyphenylacetic acid, 2-2-bromo-4-methoxyphenyl acetic acid, 2-bromo-4-methoxyphenyl acetic acid, benzeneacetic acid, 2-bromo-4-methoxy, 2-bromo-4-methoxyphenylacetci acid, pubchem3983, 2-2-bromo-4-methoxy-phenyl acetic acid, benzeneacetic acid,2-bromo-4-methoxy, 2-bromo-4-methoxy-phenyl-acetic acid
PubChem CID 3963668
IUPAC Name 2-(2-bromo-4-methoxyphenyl)acetic acid
SMILES COC1=CC(Br)=C(CC(O)=O)C=C1

Specifications

Melting Point 127°C to 131°C
Quantity 1 g
Formula Weight 245.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-methoxyphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.