missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-nitro-1-(trifluoromethoxy)benzene, 98%

Product Code. 11336559
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336559 1 g 1g
11346559 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336559 Supplier Thermo Scientific Alfa Aesar Supplier No. H26173.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 200958-40-3 | C7H3BrF3NO3 | 286.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200958-40-3
Molecular Formula C7H3BrF3NO3
Molecular Weight (g/mol) 286.00
MDL Number MFCD04973758
InChI Key LVTAFGFYMLODQP-UHFFFAOYSA-N
PubChem CID 24721641
IUPAC Name 2-bromo-4-nitro-1-(trifluoromethoxy)benzene
SMILES [O-][N+](=O)C1=CC(Br)=C(OC(F)(F)F)C=C1

Specifications

Refractive Index 1.508
Quantity 1 g
Synonym 2-bromo-4-nitro-1-trifluoromethoxy benzene, 2-bromo-4-nitro trifluoromethoxy benzene, benzene, 2-bromo-4-nitro-1-trifluoromethoxy, acmc-20ahjp, ksc496o8t, 2-bromo-4-nitro trifluomethoxy benzene, benzene,2-bromo-4-nitro-1-trifluoromethoxy, 2-bromanyl-4-nitro-1-trifluoromethyloxy benzene
Formula Weight 286
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-nitro-1-(trifluoromethoxy)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.