missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethyl)aniline, 95%, Maybridge

Product Code. 10149272
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10149272 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10149272 Supplier Thermo Scientific Maybridge Supplier No. SEW01985DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 57946-63-1
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C7H5BrF3N
MDL Number MFCD00042150
Quantity 1 g
UN Number 2811
Synonym 2-bromo-4-trifluoromethyl aniline, 4-amino-3-bromobenzotrifluoride, 2-bromo-4-trifluoromethyl-phenylamine, benzenamine, 2-bromo-4-trifluoromethyl, 3-bromo-4-aminobenzotrifluoride, 4-amino-3-bromo benzotrifluoride, buttpark 83\07-26, timtec-bb sbb000789, buttpark 44\03-51, 2-bromo-4-trifluoromethyl phenylamine
InChI Key QKRJIXSZTKOFTD-UHFFFAOYSA-N
SMILES NC1=CC=C(C=C1Br)C(F)(F)F
IUPAC Name 2-bromo-4-(trifluoromethyl)aniline
Molecular Weight (g/mol) 240.02
PubChem CID 93899
Formula Weight 240.02
Percent Purity 95%
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)aniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.