missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethyl)aniline, 97%

Product Code. 11498437
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498437 1 g 1g
11408447 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498437 Supplier Thermo Scientific Alfa Aesar Supplier No. A18528.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 57946-63-1 | C7H5BrF3N | 240.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57946-63-1
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
MDL Number MFCD00042150
InChI Key QKRJIXSZTKOFTD-UHFFFAOYSA-N
Synonym 2-bromo-4-trifluoromethyl aniline, 4-amino-3-bromobenzotrifluoride, 2-bromo-4-trifluoromethyl-phenylamine, benzenamine, 2-bromo-4-trifluoromethyl, 3-bromo-4-aminobenzotrifluoride, 4-amino-3-bromo benzotrifluoride, buttpark 83\07-26, timtec-bb sbb000789, buttpark 44\03-51, 2-bromo-4-trifluoromethyl phenylamine
PubChem CID 93899
IUPAC Name 2-bromo-4-(trifluoromethyl)aniline
SMILES NC1=CC=C(C=C1Br)C(F)(F)F

Specifications

Melting Point 26°C to 33°C
Density 1.7
Boiling Point 109°C to 110°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.524
Quantity 1 g
UN Number UN2811
Beilstein 8139978
Formula Weight 240.03
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.