missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethyl)phenylacetonitrile, 98%

Product Code. 15460428
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460428 5 g 5g
15450428 1 g 1g
15440656 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460428 Supplier Thermo Scientific Alfa Aesar Supplier No. H53518.06
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 474024-36-7 | C9H5BrF3N | 264.05 g/mol

2-Bromo-4-(trifluoromethyl)phenylacetonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-(trifluoromethyl)phenylacetonitrile is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 474024-36-7
Molecular Formula C9H5BrF3N
Molecular Weight (g/mol) 264.05
MDL Number MFCD03094308
InChI Key RYUZTTSDBHOIAF-UHFFFAOYSA-N
PubChem CID 2778436
IUPAC Name 2-[2-bromo-4-(trifluoromethyl)phenyl]acetonitrile
SMILES FC(F)(F)C1=CC(Br)=C(CC#N)C=C1

Specifications

Density 1.626
Boiling Point 265°C to 266°C
Refractive Index 1.502
Quantity 5 g
Synonym 2-bromo-4-trifluoromethyl phenylacetonitrile, 2-2-bromo-4-trifluoromethyl phenyl acetonitrile, 2-2-bromo-4-trifluoromethyl phenyl ethanenitrile, pubchem4248, 2-bromo-4-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile,2-bromo-4-trifluoromethyl, benzeneacetonitrile, 2-bromo-4-trifluoromethyl
Solubility Information Slightly soluble in water.
Formula Weight 264.04
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)phenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.