missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluoroaniline, 98%

Product Code. 11481367
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481367 5 g 5g
11491367 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481367 Supplier Thermo Scientific Alfa Aesar Supplier No. B24901.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1003-99-2 | C6H5BrFN | 190.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-99-2
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD00070750
InChI Key FWTXFEKVHSFTDQ-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-5-fluoro, 2-bromo-5-fluorobenzenamine, 2-bromo-5-fluoro-aniline, 2-bromo-5-fluoro-phenylamine, 2-bromo-5-fluoro-benzenamine, 2-bromo-5-fluorophenylamine, pubchem1524, 5-fluoro-2-bromoaniline, 2-bromo-5-fluoro aniline, acmc-1c3a5
PubChem CID 2773317
IUPAC Name 2-bromo-5-fluoroaniline
SMILES C1=CC(=C(C=C1F)N)Br

Especificaciones

Melting Point 42°C to 46°C
Boiling Point 154°C (23 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2811
Formula Weight 190.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-fluoroaniline

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.