missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzaldehyde, 98%

Product Code. 11456057
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11456057

Brand: Thermo Scientific Alfa Aesar B25071.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94569-84-3 | C7H4BrFO | 203.01 g/mol

2-Bromo-5-fluorobenzaldehyde is used as a precursor for the synthesis of 5-fluoro-3-substituted benzoxaboroles, which is used in material science as molecular receptors, building block in crystal engineering, as steroid conjugates for molecular imprinting , dyes and biosensors of alpha hydroxyl carboxylic acids. It is also used in the synthesis of 5-arylindazolo[3,2-b]quinazolin-7(5H)-one by reacting with 2-amino-N'-arylbenzohydrazide in the presence of Copper(I) bromide by the Ullmann-type reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-fluorobenzaldehyde is used as a precursor for the synthesis of 5-fluoro-3-substituted benzoxaboroles, which is used in material science as molecular receptors, building block in crystal engineering, as steroid conjugates for molecular imprinting , dyes and biosensors of alpha hydroxyl carboxylic acids. It is also used in the synthesis of 5-arylindazolo[3,2-b]quinazolin-7(5H)-one by reacting with 2-amino-N′-arylbenzohydrazide in the presence of Copper(I) bromide by the Ullmann-type reaction.

Solubility
Soluble in methanol.

Notes
Air sensitive. Keep container tightly closed in a cool, dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94569-84-3
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00142872
InChI Key CJUCIKJLMFVWIS-UHFFFAOYSA-N
Synonym benzaldehyde, 2-bromo-5-fluoro, 2-bromo-5-fluorobenzaldhyde, 5-fluoro-2-bromo benzaldehyde, 2-bromo-5-fluoro-benzaldehyde, pubchem1418, acmc-209rsk, ksc494g0d, 2-bromo-5-fluoro benzaldehyde, 2-bromo-5-fluorobenzaldehyde
PubChem CID 2773321
IUPAC Name 2-bromo-5-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1F)C=O)Br

Specifications

Melting Point 51°C to 56°C
Flash Point 87°C (190°F)
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Formula Weight 203.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-fluorobenzaldehyde
compliance-icons
Product Identifier
  • 2-Bromo-5-fluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Bromo-5-fluorobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.