missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxybenzoic acid, 97%, Thermo Scientific™

Product Code. 10587814 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10587814 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10587814 Supplier Thermo Scientific Acros Supplier No. 398930050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 22921-68-2
Melting Point 158.0°C to 160.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H7BrO3
Quantity 5 g
Synonym 6-bromo-m-anisic acid, 2-bromo-5-methoxybenzoicacid, benzoic acid, 2-bromo-5-methoxy, 5-methoxy-2-bromobenzoic acid, 2-bromo-5-methoxy-benzoic acid, pubchem2674, acmc-1clk6, ksc205k0l, rarechem al be 1111, 2-bromo-5-methoxy benzoic acid
InChI Key ODHJOROUCITYNF-UHFFFAOYSA-N
SMILES COC1=CC(=C(C=C1)Br)C(=O)O
IUPAC Name 2-bromo-5-methoxybenzoic acid
Molecular Weight (g/mol) 231.04
PubChem CID 89906
Formula Weight 231.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-methoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-5-methoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.