missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxybenzoic acid, 98+%

Product Code. p-7082029
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11474963

Brand: Thermo Scientific Alfa Aesar A12261.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22921-68-2 | C8H7BrO3 | 231.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22921-68-2
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD00020214
InChI Key ODHJOROUCITYNF-UHFFFAOYSA-N
Synonym 6-bromo-m-anisic acid, 2-bromo-5-methoxybenzoicacid, benzoic acid, 2-bromo-5-methoxy, 5-methoxy-2-bromobenzoic acid, 2-bromo-5-methoxy-benzoic acid, pubchem2674, acmc-1clk6, ksc205k0l, rarechem al be 1111, 2-bromo-5-methoxy benzoic acid
PubChem CID 89906
IUPAC Name 2-bromo-5-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)Br)C(=O)O

Specifications

Melting Point 156°C to 160°C
Odor Odorless
Quantity 10 g
Beilstein 639763
Formula Weight 231.05
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-methoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.