missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11420110
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420110 1 g 1g
11410110 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420110 Supplier Thermo Scientific Alfa Aesar Supplier No. H50608.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138642-47-4
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.046
MDL Number MFCD01861260
InChI Key ZQONVYONOASKIY-UHFFFAOYSA-N
Synonym benzonitrile,2-bromo-5-methoxy, 2-bromo-5-methoxybenzenecarbonitrile, 4-bromo-3-cyanoanisole, acmc-20ag98, 2-bromo-5-methoxy benzonitrile, 2-bromo-5-methoxybenzonitrile, 2-bromanyl-5-methoxy-benzenecarbonitrile
PubChem CID 21784096
IUPAC Name 2-bromo-5-methoxybenzonitrile
SMILES COC1=CC(=C(C=C1)Br)C#N

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 212.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-methoxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.