missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylbenzotrifluoride, 98%, Thermo Scientific Chemicals

Product Code. 15404127
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404127 1 g 1g
15414127 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404127 Supplier Thermo Scientific Alfa Aesar Supplier No. H31523.03
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 261952-20-9 | Molecular Formula: C8H6BrF3 | Molecular Weight: 239.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 261952-20-9
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.035
MDL Number MFCD01631586
InChI Key WXXDBPIOFOYRLP-UHFFFAOYSA-N
Synonym 2-bromo-5-methylbenzotrifluoride, 1-bromo-4-methyl-2-trifluoromethyl benzene, 4-bromo-3-trifluoromethyl toluene, 4-bromo-alpha,alpha,alpha-trifluoro-m-xylene, 4-methyl-2-trifluoromethyl bromobenzene, pubchem23748, 4-bromo-3-trifluoromethyltoluene, 1-bromo-4-methyl-2-trifluoromethyl-benzene, 1-bromanyl-4-methyl-2-trifluoromethyl benzene, benzene,1-bromo-4-methyl-2-trifluoromethyl
PubChem CID 2779339
IUPAC Name 1-bromo-4-methyl-2-(trifluoromethyl)benzene
SMILES CC1=CC(=C(C=C1)Br)C(F)(F)F

Especificaciones

Quantity 1 g
Formula Weight 239.04
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-methylbenzotrifluoride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.