missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitroanisole, 98%

Product Code. 11477006
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477006 2 g 2g
11487006 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477006 Supplier Thermo Scientific Alfa Aesar Supplier No. B25331.04
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 77337-82-7 | C7H6BrNO3 | 232.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77337-82-7
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.033
MDL Number MFCD00041250
InChI Key NTKADLOYTKVXQN-UHFFFAOYSA-N
Synonym 2-bromo-5-nitroanisole, 4-bromo-3-methoxynitrobenzene, 2-bromo-5-nitroanisol, benzene, 1-bromo-2-methoxy-4-nitro, 1-bromo-2-methoxy-4-nitro-benzene, zlchem 673, pubchem2536, 2-bromo 5-nitro anisole, 2-bromo-5-nitro-anisole, 2-bromo-5-nitro anisole
PubChem CID 101293
IUPAC Name 1-bromo-2-methoxy-4-nitrobenzene
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 102°C to 107°C
Quantity 2 g
Beilstein 2520496
Formula Weight 232.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-nitroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.