missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrobenzoic acid, Thermo Scientific™

Product Code. 10387601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10387601 5 g Glass bottle 5g
10123341 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10387601 Supplier Thermo Scientific Acros Supplier No. 342290050
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 943-14-6
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.02
MDL Number MFCD00134558
InChI Key UVFWYVCDRKRAJH-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrobenzoic acid, 2-bromo-5-nitrobenzic acid, benzoic acid, 2-bromo-5-nitro, 2-bromo-5-nitrobenzoicacid, 2-bromo-5-nitro benzoic acid, pubchem3821, acmc-209rrc, ksc494m1r, paragos 440009, timtec-bb sbb003179
PubChem CID 243025
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Br

Specifications

Melting Point 180°C to 181°C
Color Beige to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 246.02
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 2-Bromo-5-nitrobenzoic acid
compliance-icons
Product Identifier
  • 2-Bromo-5-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.