missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluorobenzaldehyde, 98%

Product Code. 15431999
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431999 25 g 25g
15421999 5 g 5g
15441999 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431999 Supplier Thermo Scientific Alfa Aesar Supplier No. H64689.14
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 360575-28-6 | C7H4BrFO | 203.01 g/mol

It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 360575-28-6
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD03407341
InChI Key PJNILWKRAKKEQM-UHFFFAOYSA-N
Synonym 6-bromo-2-fluorobenzaldehyde, 2-bromo-6-fluorobenzenecarbaldehyde, 2-bromo-6-fluoro-benzaldehyde, 2-fluoro-6-bromobenzaldehyde, 6-fluoro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-6-fluoro, pubchem1419, acmc-1afpf, ksc497k8t
PubChem CID 22473977
IUPAC Name 2-bromo-6-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Br)C=O)F

Specifications

Melting Point 43°C to 47°C
Density 1.70 g/mL
Boiling Point 100°C to 102°C (8 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2573976
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 203.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-fluorobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-6-fluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromo-6-fluorobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.