missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-N-methylaniline, 95%

Product Code. 15411328
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411328 1 g 1g
15431328 250 mg 250mg
15421328 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411328 Supplier Thermo Scientific Alfa Aesar Supplier No. H55166.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6832-87-7 | C7H8BrN | 186.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6832-87-7
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD06798064
InChI Key SMVIAQFTVWDWDS-UHFFFAOYSA-N
PubChem CID 334086
IUPAC Name 2-bromo-N-methylaniline
SMILES CNC1=CC=CC=C1Br

Specifications

Color Brown
Density 1.589 g/mL
Boiling Point 107°C to 109°C (12 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.607
Quantity 1 g
Synonym 2-bromo-n-methylbenzenamine, acmc-20abqa, o-bromo-n-methylaniline, n-methyl-2-bromoaniline, benzenamine,2-bromo-n-methyl, 2-bromo-n-methylaniline, benzenamine, 2-bromo-n-methyl
Formula Weight 186.05
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Bromo-N-methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.