missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-N,N-diethylethylamine hydrobromide, 98%

Product Code. 10061554 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10061554

Brand: Thermo Scientific Acros 426860250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1069-72-3 | C6H15Br2N | 261.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1069-72-3
Molecular Formula C6H15Br2N
Molecular Weight (g/mol) 261.00
MDL Number MFCD00040376
InChI Key HLMHCDKXKXBKQK-UHFFFAOYSA-N
Synonym 2-diethylamino ethyl bromide hydrobromide, 2-bromo-n,n-diethylethylamine hydrobromide, 2-bromoethyldiethylammonium bromide, 2-bromoethyl diethylamine hydrobromide, 2-bromo-n,n-diethylethanaminium bromide, 2-bromotriethylamine hydrobromide, 2-bromoethyldiethylamine hydrobromide, diethylaminoethyl bromide hydrobromide, 2-bromo-n,n-diethylethanamine hydrobromide, ethanamine, 2-bromo-n,n-diethyl-, hydrobromide
PubChem CID 70607
IUPAC Name 2-bromo-N,N-diethylethanamine;hydrobromide
SMILES [H+].[Br-].CCN(CC)CCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 211°C to 215°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3CH2)2N(CH2)2Br·HBr
Quantity 25 g
Formula Weight 260.99
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-N, N-diethylethylamine hydrobromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-N,N-diethylethylamine hydrobromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.