missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoaniline, 98%

Product Code. 11340260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11340260 500 g Glass bottle 500g
10549980 25 g Glass bottle 25g
10325690 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11340260 Supplier Thermo Scientific Acros Supplier No. 106615000
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 615-36-1 | C6H6BrN | 172.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-36-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.02
InChI Key AOPBDRUWRLBSDB-UHFFFAOYSA-N
PubChem CID 11992
IUPAC Name 2-bromoaniline
SMILES C1=CC=C(C(=C1)N)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29.0°C to 31.0°C
Density 1.5780g/mL
Boiling Point 229.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4NH2
Refractive Index 1.6170 to 1.6190
Quantity 500 g
Beilstein 12,631
Specific Gravity 1.578
Synonym o-bromoaniline, benzenamine, 2-bromo, o-aminobromobenzene, aniline, o-bromo, 2-bromobenzenamine, bromoaniline 2-, 2-bromo aniline, 2-bromophenylamine, benzenamine,2-bromo, 2-bromioaniline
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,alcohols
Formula Weight 172.02
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 2-Bromoaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Bromoaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.