missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoaniline, 98%

Product Code. 11479363
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479363 25 g 25g
11489363 100 g 100g
11499363 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479363 Supplier Thermo Scientific Alfa Aesar Supplier No. A11788.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 615-36-1 | C6H6BrN | 172.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Bromoaniline is used as a corrosion inhibitor, emulsifying and antiseptic agents. It is used in the rubber industry, e.g. diphenylguanidines, phenylenediamines mercaptobenzothiazoles, aniline ketones and etc.

Solubility
Insoluble in water.

Notes
Store at 2-8°C. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-36-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.025
MDL Number MFCD00007632
InChI Key AOPBDRUWRLBSDB-UHFFFAOYSA-N
Synonym o-bromoaniline, benzenamine, 2-bromo, o-aminobromobenzene, aniline, o-bromo, 2-bromobenzenamine, bromoaniline 2-, 2-bromo aniline, 2-bromophenylamine, benzenamine,2-bromo, 2-bromioaniline
PubChem CID 11992
IUPAC Name 2-bromoaniline
SMILES C1=CC=C(C(=C1)N)Br

Specifications

Melting Point 27°C to 31°C
Density 1.578
Boiling Point 228°C to 230°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 25 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 172.03
Percent Purity 98%
Chemical Name or Material 2-Bromoaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.