missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoanisole, 98%

Product Code. 11499193
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499193 500 g 500g
11479193 25 g 25g
11489193 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499193 Supplier Thermo Scientific Alfa Aesar Supplier No. A11692.36
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 578-57-4 | C7H7BrO | 187.036 g/mol

2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.

Solubility
Soluble in ethanol and diethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 578-57-4
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00000064
InChI Key HTDQSWDEWGSAMN-UHFFFAOYSA-N
Synonym 2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
PubChem CID 11358
IUPAC Name 1-bromo-2-methoxybenzene
SMILES COC1=CC=CC=C1Br

Specifications

Melting Point 2°C to 3°C
Density 1.511
Boiling Point 220°C to 222°C
Flash Point 96°C (204°F)
Assay Percent Range 98%
Refractive Index 1.574
Quantity 500 g
Beilstein 1859996
Solubility Information Soluble in ethanol and diethyl ether.
Formula Weight 187.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromoanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.