missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzene-1-sulfonamide, 97%, Thermo Scientific™

Product Code. 10662273
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10662273 10 g 10g
10213482 1 g 1g
10413014 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10662273 Supplier Thermo Scientific Maybridge Supplier No. TL00539EA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 92748-09-9
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.08
MDL Number MFCD00173662
InChI Key YSFGGXNLZUSHHS-UHFFFAOYSA-N
Synonym 2-bromobenzene-1-sulfonamide, 2-bromobenzenesulphonamide, 2-bromo-benzenesulfonamide, 2-bromobenzene sulfonamide, 2-bromo benzene sulfon amide, buttpark 89\07-69, 2-bromobenzene-1-sulphonamide, bromobenzenesulfonamide, bromobenzene sulfonamide, pubchem13521
PubChem CID 2801263
IUPAC Name 2-bromobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=CC=C1Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula BrC6H4SO2NH2
Quantity 10 g
Formula Weight 236.09
Percent Purity 97%
Chemical Name or Material 2-Bromobenzene-1-sulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.