missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoyl chloride, 98%

Product Code. p-72745 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 10676862

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7154-66-7 | C7H4BrClO | 219.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7154-66-7
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD00000655
InChI Key NZCKTGCKFJDGFD-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-bromo, o-bromobenzoyl chloride, benzoyl chloride, o-bromo, 2-bromo-benzoyl chloride, 2-bromobenzoylchloride, 2-bromobenzene-1-carbonyl chloride, o-brombenzoylchlorid, bromo benzoylchloride, o-bromobenzoylchloride, pubchem12510
PubChem CID 23542
IUPAC Name 2-bromobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 11.0°C
Color Colorless to Yellow
Density 1.6700g/mL
Boiling Point 245.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrC6H4COCl
Refractive Index 1.5947 to 1.5967
Quantity 10 mL
Beilstein 09, 348
Specific Gravity 1.67
Solubility Information Solubility in water: decomposes in contact with water. Other solubilities: slightly soluble in carbontetrachloride
Formula Weight 219.46
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromobenzoyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.