missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoyl chloride, 98%

Product Code. 11411878
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411878 50 g 50g
11401878 10 g 10g
11421878 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11411878 Supplier Thermo Scientific Alfa Aesar Supplier No. A14520.18
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7154-66-7 | C7H4BrClO | 219.462 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7154-66-7
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
MDL Number MFCD00000655
InChI Key NZCKTGCKFJDGFD-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-bromo, o-bromobenzoyl chloride, benzoyl chloride, o-bromo, 2-bromo-benzoyl chloride, 2-bromobenzoylchloride, 2-bromobenzene-1-carbonyl chloride, o-brombenzoylchlorid, bromo benzoylchloride, o-bromobenzoylchloride, pubchem12510
PubChem CID 23542
IUPAC Name 2-bromobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)Br

Specifications

Melting Point 10°C
Density 1.674
Boiling Point 245°C
Flash Point 107°C (224°F)
Refractive Index 1.596
Quantity 50 g
UN Number UN3265
Beilstein 508506
Sensitivity Moisture sensitive
Formula Weight 219.47
Percent Purity 98%
Chemical Name or Material 2-Bromobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.