missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl mercaptan, 99%

Product Code. 15403437
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15403437

Brand: Thermo Scientific Alfa Aesar H26247.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143888-85-1 | C7H7BrS | 203.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143888-85-1
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.10
MDL Number MFCD04038925
InChI Key UJNSDLRPHRMVGZ-UHFFFAOYSA-N
Synonym 2-bromobenzyl mercaptan, 2-bromobenzylmercaptan, 2-bromophenyl methanethiol, o-bromobenzyl hydrosulfide, 2-bromobenzylthiol, 2-bromophenyl methane-1-thiol, 2-bromobenzyl, 2-bromobenzyl mercap, pubchem6847
PubChem CID 2773673
IUPAC Name (2-bromophenyl)methanethiol
SMILES SCC1=CC=CC=C1Br

Specifications

Density 1.527
Boiling Point 256°C to 257°C
Flash Point >110°C (230°F)
Refractive Index 1.617
Quantity 250 mg
UN Number UN2810
Sensitivity Air Sensitive
Formula Weight 203.1
Percent Purity 99%
Chemical Name or Material 2-Bromobenzyl mercaptan

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.