missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzylamine, 98%

Product Code. 11985241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11985241 25 g Glass bottle 25g
10282892 1 g Glass bottle 1g
10038821 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11985241 Supplier Thermo Scientific Acros Supplier No. 350510250
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3959-05-5 | C7H8BrN | 186.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3959-05-5
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00025572
InChI Key NOYASZMZIBFFNZ-UHFFFAOYSA-N
Synonym 2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo
PubChem CID 334072
IUPAC Name (2-bromophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2NH2
Refractive Index 1.5875 to 1.5910
Quantity 25 g
Beilstein 12,III,2348
Formula Weight 186.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromobenzylamine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromobenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.