missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10626442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10626442 5 g Glass bottle 5g
10296830 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10626442 Supplier Thermo Scientific Acros Supplier No. 209090050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5465-63-4
Molecular Formula C7H8BrClN
Molecular Weight (g/mol) 221.50
MDL Number MFCD00012853
InChI Key ZANQNQMPTMEAIP-UHFFFAOYSA-N
Synonym 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2
PubChem CID 2724202
IUPAC Name (2-bromophenyl)methanamine;hydrochloride
SMILES [Cl].NCC1=CC=CC=C1Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 225°C to 230°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula BrC6H4CH2NH2·HCl
Quantity 5 g
Beilstein 12, 1074
Formula Weight 222.52
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2-Bromobenzylamine hydrochloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromobenzylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.