missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzylamine hydrochloride, 97%

Product Code. 11443667
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443667 5 g 5g
11453667 25 g 25g
11463667 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443667 Supplier Thermo Scientific Alfa Aesar Supplier No. A18151.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5465-63-4 | C7H8BrClN | 221.50 g/mol

It is employed as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate in organic syntheses.

Solubility
Soluble in methanol (25 mg/ml).

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5465-63-4
Molecular Formula C7H8BrClN
Molecular Weight (g/mol) 221.50
MDL Number MFCD00012853
InChI Key ZANQNQMPTMEAIP-UHFFFAOYSA-N
Synonym 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2
PubChem CID 2724202
SMILES [Cl].NCC1=CC=CC=C1Br

Specifications

Melting Point 227°C to 230°C
Quantity 5 g
Beilstein 3913456
Sensitivity Hygroscopic
Solubility Information Soluble in methanol (25mg/ml).
IUPAC Name (2-bromophenyl)methanamine;hydrochloride
Formula Weight 222.52
Percent Purity 97%
Chemical Name or Material 2-Bromobenzylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.