missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzylmagnesium bromide, 0.25M solution in diethyl ether, AcroSeal™

Product Code. 11955891 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11955891

Brand: Thermo Scientific Acros 431611000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56812-60-3 | C7H6Br2Mg | 274.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56812-60-3, 60-29-7
CAS Min % 5.37, 87.47
CAS Max % 12.53, 94.63
Color Yellow to Brown
Density 0.7660g/mL
Boiling Point 34.6°C
Flash Point −40°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C7H6Br2Mg
MDL Number MFCD01319903
Quantity 100 mL
Specific Gravity 0.766
Synonym 2-bromobenzylmagnesium bromide, hdxzjaztpvlpex-uhfffaoysa-m, 2-bromobenzylmagnesium bromide solution, bromo 2-bromophenyl methyl magnesium, 2-bromobenzylmagnesium bromide 0.25 m in diethyl ether, 2-bromobenzylmagnesium bromide, 0.25m in diethyl ether, 2-bromobenzylmagnesium bromide solution, 0.25 m in diethyl ether, grignard reagent
InChI Key HDXZJAZTPVLPEX-UHFFFAOYSA-M
SMILES Br[Mg]CC1=CC=CC=C1Br
IUPAC Name bromo[(2-bromophenyl)methyl]magnesium
Molecular Weight (g/mol) 274.24
PubChem CID 10858676
Formula Weight 274.24
Physical Form Solution
Chemical Name or Material 2-Bromobenzylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Bromobenzylmagnesium bromide, 0.25M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.