missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobiphenyl, 98%

Product Code. p-7026058
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11454887

Brand: Thermo Scientific Alfa Aesar B21840.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2052-07-5 | C12H9Br | 233.11 g/mol

It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-07-5
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.11
MDL Number MFCD00000065
InChI Key KTADSLDAUJLZGL-UHFFFAOYSA-N
Synonym 2-bromobiphenyl, o-bromobiphenyl, 2-bromo-biphenyl, 1,1'-biphenyl, 2-bromo, 2-bromo-1,1'-biphenyl, biphenyl, 2-bromo, 2-bromo-diphenyl, bromobiphenyl, 2-bromodiphenyl, 1,1'-biphenyl, bromo
PubChem CID 16329
IUPAC Name 1-bromo-2-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2Br

Specifications

Melting Point 0°C to 2°C
Density 1.376
Boiling Point 298°C
Flash Point >110°C (230°F)
Refractive Index 1.629
Quantity 1 g
Beilstein 1863156
Solubility Information Insoluble in water.
Formula Weight 233.12
Percent Purity 98%
Chemical Name or Material 2-Bromobiphenyl
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.