missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutyric acid, 99%

Product Code. p-72796 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
5mL
This item is not returnable. View return policy

Product Code. 10580702

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-58-0 | C4H7BrO2 | 167.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-58-0
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00004216,MFCD00210111,MFCD00210112
InChI Key YAQLSKVCTLCIIE-UHFFFAOYNA-N
Synonym 2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid
PubChem CID 6655
IUPAC Name 2-bromobutanoic acid
SMILES CCC(Br)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -4°C
Color Yellow
Density 1.5600g/mL
Boiling Point 214°C to 217°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C2H5CH(Br)CO2H
Refractive Index 1.4712 to 1.4732
Quantity 5 mL
Beilstein 02, 281
Merck Index 15, 1420
Specific Gravity 1.56
Solubility Information Solubility in water: 66g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 167
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromobutyric acid, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromobutyric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.