missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethanesulfonic acid, sodium salt, 98%

Produktkod. 10581981 Handla allt Thermo Scientific Chemicals Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
25 g
100 g
Förpackningsstorlek:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
10581981 25 g 25g
10784661 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10581981 Leverantör Thermo Scientific Acros Leverantörsnummer 160030250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 4263-52-9 | C2H4BrNaO3S | 211.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 4263-52-9
Molecular Formula C2H4BrNaO3S
Molecular Weight (g/mol) 211.01
MDL Number MFCD00007530
InChI Key HNFOAHXBHLWKNF-UHFFFAOYSA-M
Synonym sodium 2-bromoethanesulfonate, 2-bromoethanesulfonic acid sodium salt, sodium 2-bromoethanesulphonate, 2-bromoethanesulfonic acid, sodium salt, sodium bromoethylsulfonate, ethanesulfonic acid, 2-bromo-, sodium salt, ethanesulfonic acid, 2-bromo-, sodium salt 1:1, sodium 2-bromoethane sulfonate, 2-bromoethanesulfonic acid sodium, natrium-2-bromethansulfonat
PubChem CID 23666797
IUPAC Name sodium;2-bromoethanesulfonate
SMILES C(CBr)S(=O)(=O)[O-].[Na+]

Specifikationer

CAS Min % 97.5
CAS Max % 100.0
Melting Point 282.0°C to 286.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Br) On dry substance (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2CH2SO3Na
Quantity 25 g
Beilstein 04, 7
Solubility Information Solubility in water: in water: 500g/L (20°C)
Formula Weight 211.01
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoethanesulfonic acid, sodium salt, 98%
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.