missing translation for 'onlineSavingsMsg'
Learn More

2-Bromofluorobenzene, 99%

Product Code. 10305790 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10305790 10 mL Glass bottle 10mL
10427290 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10305790 Supplier Thermo Scientific Supplier No. 106990100
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1072-85-1 | C6H4BrF | 175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-85-1
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175
MDL Number MFCD00000282
InChI Key IPWBFGUBXWMIPR-UHFFFAOYSA-N
Synonym 2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
PubChem CID 61259
IUPAC Name 1-bromo-2-fluorobenzene
SMILES C1=CC=C(C(=C1)F)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6000g/mL
Boiling Point 155.0°C to 157.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4F
Refractive Index 1.5327 to 1.5347
Quantity 10 mL
Fieser 01,77
Specific Gravity 1.6
Solubility Information Solubility in water: insoluble in water
Formula Weight 175
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromofluorobenzene, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.