missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisophthalaldehyde, 97%

Product Code. 15421629
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421629 5 g 5g
15431629 250 mg 250mg
15411629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421629 Supplier Thermo Scientific Alfa Aesar Supplier No. H64423.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79839-49-9 | C8H5BrO2 | 213.03 g/mol

Used in the preparation of paclitaxel-mimicking alkaloids with anticancer activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of paclitaxel-mimicking alkaloids with anticancer activity.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79839-49-9
Molecular Formula C8H5BrO2
Molecular Weight (g/mol) 213.03
MDL Number MFCD08461034
InChI Key RZUSSKMZLHKMHU-UHFFFAOYSA-N
Synonym 2-bromoisophthalaldehyde, 2-bromobenzene-1,3-dialdehyde, 2-bromo-1,3-diformylbenzene, 2,6-diformylbromobenzene, 2-bromo-benzene-1,3-dicarbaldehyde, 2-bromobenzene-1,3-dicarboxaldehyde, 1,3-diformyl-2-bromobenzene, 1,3-benzenedicarboxaldehyde, 2-bromo, acmc-209pia, ksc495c0d
PubChem CID 11435812
IUPAC Name 2-bromobenzene-1,3-dicarbaldehyde
SMILES BrC1=C(C=O)C=CC=C1C=O

Specifications

Melting Point 139°C to 144°C
Quantity 5 g
Beilstein 4740331
Solubility Information Slightly soluble in water.
Formula Weight 213.03
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromoisophthalaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.